siRNA Details
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037978.1 | NP_001033067.1 | 4 | 483 | ||
XM_006255083.1 | XP_006255145.1 | 4 | 569 | |||
GenBank mRNA | BC099768.1 | - | 4 | 483 | 7932140 |
