siRNA Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037196.1 | NP_001032273.1 | 6, 7 | 1268 | ||
XM_006230258.2 | XP_006230320.1 | 6, 7 | 1955 | |||
GenBank mRNA | BC100627.1 | - | 6, 7 | 1268 | 7938034 |
