siRNA Details
Gene Symbol: Adamtsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034012.1 | NP_001029184.1 | 16 | 3297 | ||
XM_008761305.1 | XP_008759527.1 | 13 | 3054 | |||
XM_008761306.1 | XP_008759528.1 | 15 | 3002 | |||
XM_008761307.1 | XP_008759529.1 | 14 | 2955 | |||
GenBank mRNA | BC085819.1 | - | 16 | 2869 | ||
BC099119.1 | - | 16 | 3297 | 7462597 |
