siRNA Details
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029919.2 | NP_001025090.1 | 8 | 1168 | ||
XM_006237305.1 | XP_006237367.1 | 9 | 1229 | |||
GenBank mRNA | BC097496.1 | - | 8 | 1167 | 7461823 | |
CO402572.1 | - | 1 | 285 |
