siRNA Details
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034149.1 | NP_001029321.1 | 7, 8 | 1166 | ||
XM_006239305.2 | XP_006239367.1 | 7, 8 | 1196 | |||
XM_006239306.2 | XP_006239368.1 | 7, 8 | 1498 | |||
XM_006239309.2 | XP_006239371.1 | 7, 8 | 1196 | |||
GenBank mRNA | BC097361.1 | - | 7, 8 | 1166 | 7442573 |
