siRNA Details
Gene Symbol: Ncapd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034000.1 | NP_001029172.1 | 13, 14 | 1712 | ||
XM_006242735.2 | XP_006242797.1 | 13, 14 | 2014 | |||
XM_006242736.2 | XP_006242798.1 | 13, 14 | 1665 | |||
XM_008766053.1 | XP_008764275.1 | 12, 13 | 2089 | |||
GenBank mRNA | BC101928.1 | - | 13, 14 | 1712 | 7939116 |
