siRNA Details
Gene Symbol: Ptprk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029902.2 | NP_001025073.2 | 3 | 474 | ||
XM_008758616.1 | XP_008756838.1 | 3 | 671 | |||
XM_008758617.1 | XP_008756839.1 | 3 | 670 | |||
XM_008758618.1 | XP_008756840.1 | 3 | 670 | |||
XM_008758619.1 | XP_008756841.1 | 3 | 678 | |||
XM_008758620.1 | XP_008756842.1 | 3 | 678 | |||
GenBank mRNA | AB297790.1 | - | 3 | 474 |
