siRNA Details
Gene Symbol: Atg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038495.1 | NP_001033584.1 | 4 | 544 | ||
GenBank mRNA | BC087139.1 | - | 4 | 527 | ||
BC110056.1 | - | 4 | 544 | 7443799 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038495.1 | NP_001033584.1 | 4 | 544 | ||
GenBank mRNA | BC087139.1 | - | 4 | 527 | ||
BC110056.1 | - | 4 | 544 | 7443799 |