siRNA Details
Gene Symbol: ARNTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248002.1 | NP_001234931.1 | 16 | 6104 | ||
NM_001248003.1 | NP_001234932.1 | 15 | 6035 | |||
NM_001248004.1 | NP_001234933.1 | 15 | 6002 | |||
NM_001248005.1 | NP_001234934.1 | 14 | 5926 | |||
NM_020183.4 | NP_064568.3 | 17 | 6146 | |||
XM_006719112.3 | XP_006719175.1 | 16 | 6105 | |||
XM_006719114.3 | XP_006719177.1 | 16 | 5836 | |||
XM_011520766.2 | XP_011519068.1 | 17 | 6147 | |||
XM_011520768.2 | XP_011519070.1 | 16 | 6055 | |||
XM_011520769.2 | XP_011519071.1 | 16 | 6045 | |||
XM_011520770.2 | XP_011519072.1 | 17 | 5965 | |||
XM_011520771.2 | XP_011519073.1 | 17 | 5931 | |||
XM_017019666.1 | XP_016875155.1 | 14 | 5876 | |||
XM_017019667.1 | XP_016875156.1 | 17 | 6163 | |||
XM_017019668.1 | XP_016875157.1 | 17 | 5960 | |||
XM_017019669.1 | XP_016875158.1 | 16 | 5807 | |||
GenBank mRNA | AF256215.1 | - | 6040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
