siRNA Details
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034110.1 | NP_001029282.1 | 14 | 1607 | ||
GenBank mRNA | BC099780.1 | - | 14 | 1607 | 7933625 | |
EU253481.1 | - | 14 | 1637 | |||
FQ222728.1 | - | 1 | 223 |
