siRNA Details
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033984.1 | NP_001029156.1 | 35 | 4312 | ||
NM_031805.1 | NP_113993.1 | 33 | 4547 | |||
GenBank mRNA | AF069525.1 | - | 33 | 4369 | ||
AF102552.1 | - | 35 | 4312 | |||
AJ428573.1 | - | 12 | 1730 | |||
AJ812019.1 | - | 12 | 2090 | |||
AJ812020.1 | - | 12 | 2090 | |||
AJ812021.1 | - | 12 | 2090 | |||
AJ812022.1 | - | 12 | 2090 | |||
AJ812023.1 | - | 33 | 4539 | |||
AJ812024.1 | - | 33 | 4539 | |||
BC101885.1 | - | 12 | 1771 |
