siRNA Details
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029922.1 | NP_001025093.1 | 5, 6 | 523 | ||
XM_006232495.2 | XP_006232557.1 | 5, 6 | 439 | |||
XM_006232496.2 | XP_006232558.1 | 5, 6 | 438 | |||
XM_008761103.1 | XP_008759325.1 | 7, 8 | 886 | |||
XM_008761104.1 | XP_008759326.1 | 7, 8 | 885 | |||
XM_008761105.1 | XP_008759327.1 | 7, 8 | 889 | |||
GenBank mRNA | BC097393.1 | - | 5, 6 | 523 | 7453249 |
