siRNA Details
Gene Symbol: Zbbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029922.1 | NP_001025093.1 | 12 | 1315 | ||
XM_006232495.2 | XP_006232557.1 | 12 | 1246 | |||
XM_006232496.2 | XP_006232558.1 | 12 | 1245 | |||
XM_008761103.1 | XP_008759325.1 | 14 | 1693 | |||
XM_008761104.1 | XP_008759326.1 | 14 | 1677 | |||
XM_008761105.1 | XP_008759327.1 | 14 | 1696 | |||
GenBank mRNA | BC097393.1 | - | 12 | 1315 | 7453249 |
