siRNA Details
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033707.1 | NP_001028879.1 | 2 | 169 | ||
XM_006234529.2 | XP_006234591.1 | 2 | 274 | |||
XM_006234530.2 | XP_006234592.1 | 2 | 273 | |||
XM_006234531.2 | XP_006234593.1 | 2 | 273 | |||
GenBank mRNA | BC101917.1 | - | 2 | 169 | 7935829 | |
FQ218185.1 | - | 2 | 241 | |||
FQ228393.1 | - | 2 | 236 |
