siRNA Details
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033707.1 | NP_001028879.1 | 4, 5 | 443 | ||
XM_006234529.2 | XP_006234591.1 | 4, 5 | 548 | |||
XM_006234530.2 | XP_006234592.1 | 4, 5 | 547 | |||
XM_006234531.2 | XP_006234593.1 | 4, 5 | 547 | |||
GenBank mRNA | BC101917.1 | - | 4, 5 | 443 | 7935829 | |
FQ218185.1 | - | 4, 5 | 515 | |||
FQ228393.1 | - | 4, 5 | 510 |
