siRNA Details
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034941.1 | NP_001030113.1 | 9 | 1212 | ||
XM_006242428.2 | XP_006242490.1 | 9 | 1292 | |||
XM_006242429.2 | XP_006242491.1 | 8 | 1105 | |||
XM_006242430.2 | XP_006242492.1 | 7 | 950 | |||
GenBank mRNA | BC104690.1 | - | 9 | 1212 | 7107060 | |
FQ228813.1 | - | 9 | 1209 |
