siRNA Details
Gene Symbol: Defb52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037524.1 | NP_001032613.1 | 3 | 182 | ||
XM_006253368.2 | XP_006253430.1 | 3 | 238 | |||
GenBank mRNA | DQ012091.1 | - | 3 | 182 |
