siRNA Details
Gene Symbol: ZBTB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317990.1 | NP_001304919.1 | 3 | 4070 | ||
NM_015898.3 | NP_056982.1 | 3 | 4234 | |||
XM_005259570.4 | XP_005259627.3 | 3 | 5156 | |||
XM_005259571.4 | XP_005259628.1 | 3 | 5004 | |||
GenBank mRNA | AL390151.1 | - | 1 | 594 | ||
BC019108.1 | - | 1 | 869 | |||
BC080608.1 | - | 1 | 581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
