siRNA Details
Gene Symbol: LOC500354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037797.1 | NP_001032886.1 | 2 | 1049 | ||
XM_006237568.2 | XP_006237630.1 | 2 | 1038 | |||
XM_008763417.1 | XP_008761639.1 | 2 | 1109 | |||
GenBank mRNA | BC098929.1 | - | 2 | 1049 | 7133030 |
