siRNA Details
Gene Symbol: Cpvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029927.1 | NP_001025098.1 | 2 | 165 | ||
XM_006236511.2 | XP_006236573.2 | 2 | 641 | |||
XM_008762915.1 | XP_008761137.1 | 4 | 950 | |||
GenBank mRNA | BC097471.1 | - | 2 | 165 | 7457768 |
