siRNA Details
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034997.1 | NP_001030169.1 | 7 | 725 | ||
GenBank mRNA | BC104694.1 | - | 7 | 725 | 7110947 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_147201, BC004756, AK046142, AK218775, AK170501, and others.
Gene Ontology Categories:
