siRNA Details
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034998.2 | NP_001030170.1 | 16 | 1621 | ||
XM_006256895.2 | XP_006256957.1 | 16 | 1623 | |||
XM_006256896.2 | XP_006256958.1 | 16 | 1911 | |||
XM_006256897.2 | XP_006256959.1 | 16 | 1744 | |||
XM_006256898.2 | XP_006256960.1 | 16 | 1705 | |||
XM_006256899.2 | XP_006256961.1 | 16 | 2076 | |||
XM_006256900.2 | XP_006256962.1 | 16 | 1719 | |||
GenBank mRNA | BC085358.1 | - | 9 | 723 | ||
BC085949.1 | - | 16 | 1621 | 7132135 |
