siRNA Details
Gene Symbol: Tmem69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035001.2 | NP_001030173.1 | 3 | 789 | ||
GenBank mRNA | BC083906.1 | - | 994 | 7133962 | ||
BC161923.1 | - | 3 | 789 | 8370867 |
