siRNA Details
Gene Symbol: GSN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103560.1 | 1 | 3662 | |||
GenBank mRNA | AF220263.1 | - | 1 | 3642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
