siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 31 | 3538 | ||
AK039085.1 | - | 14 | 2901 | |||
AK046816.1 | - | 19 | 2634 | |||
AK048406.1 | - | 13 | 1416 | |||
AK081253.1 | - | 3 | 163 | |||
AK129355.1 | - | 25 | 3008 | |||
AK147625.1 | - | 29 | 5487 | |||
BC057073.1 | - | 7 | 1124 | |||
BC141190.1 | - | 30 | 3583 | 9055802 | ||
BC171963.1 | - | 30 | 3583 | 9054249 | ||
FJ628361.1 | - | 31 | 3581 |
