siRNA Details
Gene Symbol: Olfr1459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146689.1 | NP_666900.1 | 1 | 693 | ||
GenBank mRNA | BC138407.1 | - | 1 | 779 | 8861427 | |
BC138408.1 | - | 1 | 779 | 8861428 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146689.1 | NP_666900.1 | 1 | 693 | ||
GenBank mRNA | BC138407.1 | - | 1 | 779 | 8861427 | |
BC138408.1 | - | 1 | 779 | 8861428 |