siRNA Details
Gene Symbol: EPPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302861.1 | NP_001289790.1 | 1, 2 | 169 | ||
NM_020398.3 | NP_065131.1 | 1, 2 | 169 | |||
GenBank mRNA | AF286368.1 | - | 1, 2 | 121 | ||
AF286370.2 | - | 1, 2 | 121 | |||
AK126022.1 | - | 1, 2 | 100 | |||
AK311906.1 | - | 1, 2 | 168 | |||
AL118493.1 | - | 1, 2 | 123 | |||
BC003675.1 | - | 1, 2 | 144 | |||
BC044829.1 | - | 1, 2 | 132 | 5171493 | ||
BC053369.1 | - | 1, 2 | 127 | 5165509 | ||
DA821702.1 | - | 1, 2 | 144 | |||
DC395362.1 | - | 1, 2 | 170 |
Gene Symbol: EPPIN-WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198986.1 | NP_001185915.1 | 1, 2 | 169 | ||
GenBank mRNA | AK301937.1 | - | 1, 2 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
