siRNA Details
Gene Symbol: AB124611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198794.1 | NP_001185723.1 | 1 | 119 | ||
NM_206536.2 | NP_996259.2 | 1 | 119 | |||
GenBank mRNA | AB124611.1 | - | 1 | 19 | ||
AB124612.1 | - | 1 | 19 | |||
AK154442.1 | - | 1 | 106 | |||
AK162293.1 | - | 1 | 106 | |||
AK168311.1 | - | 1 | 106 | |||
AK170200.1 | - | 1 | 106 | |||
BU696592.1 | - | 1 | 120 |
