siRNA Details
Gene Symbol: Olfr1434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146685.3 | NP_666896.2 | 2 | 839 | ||
GenBank mRNA | BC132136.1 | - | 1 | 767 | 40130413 | |
BC138096.1 | - | 1 | 767 | 8861115 | ||
BC147691.1 | - | 1 | 768 | 9056664 | ||
BC147692.1 | - | 1 | 768 | 9056665 |
