siRNA Details
Gene Symbol: Zfp748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035231.3 | NP_001030308.1 | 3 | 298 | ||
GenBank mRNA | AK011397.1 | - | 3 | 284 | ||
AK132196.1 | - | 3 | 285 | |||
AK141365.1 | - | 3 | 282 | |||
AK220562.1 | - | 3 | 277 | |||
BC055310.1 | - | 3 | 284 | 30048242 | ||
CN460452.1 | - | 3 | 298 |
