siRNA Details
Gene Symbol: Lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029878.1 | NP_001025049.1 | 11 | 2053 | ||
GenBank mRNA | BC096025.1 | - | 2 | 839 | ||
BC139127.1 | - | 11 | 2104 | 8862149 | ||
BC139130.1 | - | 11 | 2104 | 8862152 |
