siRNA Details
Gene Symbol: 2610028H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029816.2 | NP_084092.2 | 6 | 669 | ||
GenBank mRNA | AK011591.1 | - | 6 | 625 | ||
AK161552.1 | - | 6 | 479 | |||
BC051454.1 | - | 6 | 606 | 6308081 | ||
BQ714071.1 | - | 6 | 688 |
