siRNA Details
Gene Symbol: Gm608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029889.2 | NP_001025060.1 | 8 | 5838 | ||
GenBank mRNA | AK173333.1 | - | 1 | 2583 | ||
BC141330.1 | - | 6 | 5627 | 9055942 | ||
BC141331.1 | - | 6 | 5627 | 9055943 |
