siRNA Details
Gene Symbol: Gmnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013761.2 | NP_001013783.1 | 6 | 2757 | ||
NM_001285916.1 | NP_001272845.1 | 5 | 2851 | |||
NM_001285918.1 | NP_001272847.1 | 5 | 2582 | |||
GenBank mRNA | BC086669.1 | - | 5 | 2512 | 30639759 |
