siRNA Details
Gene Symbol: GOPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020399.3 | NP_065132.1 | 3, 4 | 730 | ||
GenBank mRNA | AB209385.1 | - | 3, 4 | 676 | ||
AF287894.1 | - | 3, 4 | 552 | |||
BX419592.2 | - | 3, 4 | 665 | |||
KF420123.1 | - | 3, 4 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
