siRNA Details
Gene Symbol: Gm4788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029977.3 | NP_001025148.3 | 5 | 722 | ||
NM_001160303.1 | NP_001153775.1 | 5 | 722 | |||
NM_001160304.1 | NP_001153776.1 | 5 | 722 | |||
GenBank mRNA | AJ871401.1 | - | 5 | 651 | ||
AJ871402.1 | - | 5 | 651 | |||
BC141136.1 | - | 5 | 694 | 9055748 | ||
BC145444.1 | - | 5 | 694 | 9054002 | ||
M29009.1 | - | 5 | 719 |
