siRNA Details
Gene Symbol: Dos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195268.1 | NP_001182197.1 | 7 | 912 | ||
GenBank mRNA | AK035824.1 | - | 1 | 63 | ||
AK137009.1 | - | 9 | 1248 | |||
BC052075.1 | - | 8 | 966 | |||
BC078442.1 | - | 3 | 467 | |||
BC100552.1 | - | 2 | 295 | 5687195 |
