siRNA Details
Gene Symbol: Inpp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024617.3 | NP_001019788.1 | 25 | 4015 | ||
NM_001297591.1 | NP_001284520.1 | 25 | 3992 | |||
GenBank mRNA | BC064813.1 | - | 23 | 3717 | 6826283 | |
DQ296613.1 | - | 25 | 3992 |
