siRNA Details
Gene Symbol: Cass4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080820.2 | NP_001074289.1 | 5 | 1877 | ||
NM_001276422.1 | NP_001263351.1 | 5 | 1883 | |||
NM_001276423.1 | NP_001263352.1 | 5 | 1877 | |||
GenBank mRNA | AK036419.1 | - | 5 | 1851 | ||
AK080960.1 | - | 5 | 1852 | |||
AK089450.1 | - | 5 | 1878 | |||
AK170726.1 | - | 5 | 1877 | |||
BC117858.1 | - | 5 | 1771 | 40095182 | ||
BC117859.1 | - | 5 | 1770 | 40095183 | ||
BC129975.1 | - | 5 | 1768 | 40095184 | ||
BC129976.1 | - | 5 | 1776 | 40095185 |
