siRNA Details
Gene Symbol: Gm773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033423.2 | NP_001028595.1 | 2 | 115 | ||
GenBank mRNA | AK144779.1 | - | 2 | 116 | ||
AK165870.1 | - | 2 | 113 | |||
BC147685.1 | - | 1 | 46 | 9056658 | ||
BC147688.1 | - | 1 | 46 | 9056661 | ||
BC172127.1 | - | 1 | 46 | |||
BC172131.1 | - | 1 | 46 |
