siRNA Details
Gene Symbol: A630001O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041328.1 | - | 2 | 2222 | ||
AK154268.1 | - | 1 | 1121 | |||
AK172442.1 | - | 2081 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A630001O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041328.1 | - | 2 | 2222 | ||
AK154268.1 | - | 1 | 1121 | |||
AK172442.1 | - | 2081 |