siRNA Details
Gene Symbol: 1700022I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026088.3 | NP_080364.1 | 1 | 156 | ||
GenBank mRNA | AK006242.1 | - | 1 | 157 | ||
AK007056.1 | - | 1 | 116 | |||
AK132843.1 | - | 157 | ||||
AK133216.1 | - | 1 | 144 |
