siRNA Details
Gene Symbol: Ankrd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029245.3 | NP_083521.2 | 6 | 1643 | ||
GenBank mRNA | AK133099.1 | - | 6 | 1645 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029245.3 | NP_083521.2 | 6 | 1643 | ||
GenBank mRNA | AK133099.1 | - | 6 | 1645 |