siRNA Details
Gene Symbol: Wnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199083.1 | NP_001186012.1 | 10 | 4602 | ||
NM_001199084.1 | NP_001186013.1 | 9 | 4344 | |||
GenBank mRNA | BC146009.1 | - | 1 | 1280 | 40131312 | |
BC146011.1 | - | 1 | 1280 | 40131314 | ||
CN693598.1 | - | 1 | 477 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001199095, NM_001002823, XM_008763215.
Gene Ontology Categories:
