siRNA Details
Gene Symbol: Gm11992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037928.3 | NP_001033017.1 | 2 | 741 | ||
GenBank mRNA | AK154909.1 | - | 2 | 742 | ||
AK160996.1 | - | 2 | 720 | |||
BC147253.1 | - | 2 | 620 | 9056290 | ||
BC147254.1 | - | 2 | 620 | 9056286 |
