siRNA Details
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020394.4 | NP_065127.4 | 4 | 607 | ||
GenBank mRNA | CN394881.1 | - | 4 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
