siRNA Details
Gene Symbol: FAM209B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013646.3 | NP_001013668.2 | 2 | 455 | ||
XM_017027849.1 | XP_016883338.1 | 4 | 971 | |||
GenBank mRNA | BC105792.1 | - | 3 | 420 | 6618487 | |
DB452055.1 | - | 2 | 416 | |||
DB517586.1 | - | 2 | 220 |
