siRNA Details
Gene Symbol: HIST2H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289664.1 | - | 1 | 556 | ||
BC110793.1 | - | 1 | 542 | 5224812 | ||
DA042901.1 | - | 1 | 559 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST2H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289664.1 | - | 1 | 556 | ||
BC110793.1 | - | 1 | 542 | 5224812 | ||
DA042901.1 | - | 1 | 559 |