siRNA Details
Gene Symbol: ABO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020469.2 | NP_065202.2 | 6 | 324 | ||
GenBank mRNA | AF134412.1 | - | 6 | 299 | ||
AF134413.1 | - | 6 | 299 | |||
AF134415.1 | - | 6 | 298 | |||
AY727862.1 | - | 1 | 60 | |||
BC069814.1 | - | 6 | 299 | |||
BC111575.1 | - | 6 | 299 | 40069575 | ||
FR691751.1 | - | 6 | 299 | |||
FR691752.1 | - | 6 | 299 | |||
J05175.1 | - | 6 | 296 | |||
JX398330.1 | - | 6 | 299 | |||
JX398331.1 | - | 6 | 299 | |||
JX398332.1 | - | 6 | 299 | |||
JX398333.1 | - | 6 | 299 | |||
JX519569.1 | - | 6 | 299 | |||
JX519570.1 | - | 6 | 299 | |||
KT282967.1 | - | 6 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
