siRNA Details
Gene Symbol: VN1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020633.3 | NP_065684.1 | 1 | 406 | ||
GenBank mRNA | AF255342.1 | - | 1 | 153 | ||
AF302903.1 | - | 1 | 153 | |||
AK094452.1 | - | 1 | 812 | |||
BC095508.1 | - | 1 | 196 | 6971941 |
